Läs mer
(3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl-[(phenylsulfonyl)oxy]acetic Acid Ester, TRC
Produktkod. 30372111
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30372111 100 mg
1 options
Produktkod. 30372111 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE678715100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-decahydro-3a,9-propano-3aH-cyclopentacyclo-octen-8-yl [(Phenylsulphonyl)oxy]acetate
CAS 848129-54-4
Molekylformel C28 H38 O7 S
Kvantitet 100 mg
Synonym (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-decahydro-3a,9-propano-3aH-cyclopentacyclo-octen-8-yl [(Phenylsulphonyl)oxy]acetate, Tiamulin Hydrogen Fumarate Imp. P (EP)
InChI formel InChI=1S/C28H38O7S/c1-6-26(4)16-22(35-23(30)17-34-36(32,33)20-10-8-7-9-11-20)27(5)18(2)12-14-28(19(3)25(26)31)15-13-21(29)24(27)28/h6-11,18-19,22,24-25,31H,1,12-17H2,2-5H3/t18-,19+,22-,24+,25+,26-,27+,28+/m1/s1
LEDER C[C@@H]1CC[C@@]23CCC(=O)[C@H]2[C@]1(C)[C@@H](C[C@@](C)(C=C)[C@@H](O)[C@@H]3C)OC(=O)COS(=O)(=O)c4ccccc4
IUPAC-namn (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-decahydro-3a,9-propano-3aH-cyclopentacyclo-octen-8-yl [(Phenylsulfonyl)oxy]acetate
Molekylvikt (g/mol) 518.66
Formel vikt 518.2338