Läs mer
(3Beta,16Alpha)-16,17-Epoxy-3-hydroxy-pregn-5-en-20-one Cyclic 1,2-Ethanediyl Acetal, TRC
Produktkod. 30376576
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30376576 1 g
1 options
Produktkod. 30376576 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5859351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3beta,16alpha)-16,17-Epoxy-3-hydroxy-pregn-5-en-20-one Cyclic 1,2-Ethanediyl Acetal
CAS 19454-86-5
Molekylformel C23H34O4
Kvantitet 1 g
Synonym 16alpha,17-Epoxy-3beta-hydroxy-pregn-5-en-20-one, Cyclic Ethylene Acetal
InChI formel InChI=1S/C23H34O4/c1-20-8-6-15(24)12-14(20)4-5-16-17(20)7-9-21(2)18(16)13-19-23(21,27-19)22(3)25-10-11-26-22/h4,15-19,24H,5-13H2,1-3H3/t15-,16+,17-,18-,19+,20-,21-,23+/m0/s1
LEDER C[C@]12CC[C@H](O)CC1=CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3C[C@H]5O[C@@]45C6(C)OCCO6
Molekylvikt (g/mol) 374.51
Formel vikt 374.2457