Läs mer
3Beta,17-Dihydroxy-16Beta-methyl-pregn-5-en-20-one Cyclic Ethylene Acetal, TRC
Produktkod. 30372954
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30372954 500 mg
30377797 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30372954 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD453699500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3beta,17-Dihydroxy-16beta-methyl-pregn-5-en-20-one Cyclic Ethylene Acetal
CAS 115605-76-0
Molekylformel C24 H38 O4
Kvantitet 500 mg
Synonym (3beta,16beta)-3,17-Dihydroxy-16-methyl-Pregn-5-en-20-one Cyclic 1,2-Ethanediyl Acetal
InChI formel InChI=1S/C24H38O4/c1-15-13-20-18-6-5-16-14-17(25)7-9-21(16,2)19(18)8-10-22(20,3)24(15,26)23(4)27-11-12-28-23/h5,15,17-20,25-26H,6-14H2,1-4H3/t15-,17-,18+,19-,20-,21-,22-,24+/m0/s1
LEDER C[C@H]1C[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@@]1(O)C5(C)OCCO5
IUPAC-namn (3S,8R,9S,10R,13S,14S,16S,17R)-10,13,16-trimethyl-17-(2-methyl-1,3-dioxolan-2-yl)-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvikt (g/mol) 390.556
Formel vikt 390.277