Läs mer
(3Beta,20S)-Pregn-5-ene-3,17,20-triol, TRC
Produktkod. 30416080
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416080 50 mg
30410123 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30416080 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP71232550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 903-67-3
Molekylformel C21H34O3
Molekylvikt (g/mol) 334.49
Synonym Pregn-5-ene-3beta,17,20alpha-triol, (3beta,17alpha,20S)-Pregn-5-ene-3,17,20-triol, Pregn-5-ene-3beta,17alpha,20alpha-triol, delta5-Pregnene-3beta,17alpha,20(S)-triol
IUPAC-namn (3S,8R,9S,10R,13S,14S,17R)-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-diol
LEDER C[C@H](O)[C@@]1(O)CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material (3beta,20S)-Pregn-5-ene-3,17,20-triol
Kvantitet 50 mg
InChI formel InChI=1S/C21H34O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h4,13,15-18,22-24H,5-12H2,1-3H3/t13-,15-,16+,17-,18-,19-,20-,21-/m0/s1
Formel vikt 334.2508