Läs mer
(3Beta,5Alpha,6Alpha,20R)-Pregnane-3,6,20-triol, TRC
Produktkod. 30416676
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416676 25 mg
30409038 10 mg
30409405 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30416676 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP03099025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70286-67-8
Molekylformel C21H36O3
Molekylvikt (g/mol) 336.51
Synonym 5a-Pregane-3b,6a,20b-triol
IUPAC-namn (3S,5S,6S,8S,9S,10R,13S,14S,17S)-17-[(1R)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,6-diol
LEDER C[C@@H](O)[C@H]1CC[C@H]2[C@@H]3C[C@H](O)[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material (3beta,5alpha,6alpha,20R)-Pregnane-3,6,20-triol
Kvantitet 25 mg
InChI formel InChI=1S/C21H36O3/c1-12(22)15-4-5-16-14-11-19(24)18-10-13(23)6-8-21(18,3)17(14)7-9-20(15,16)2/h12-19,22-24H,4-11H2,1-3H3/t12-,13+,14+,15-,16+,17+,18-,19+,20-,21-/m1/s1
Formel vikt 336.2664