Läs mer
(3Beta,5Alpha)-Cholesta-8,14-dien-3-ol, TRC
Produktkod. 30360799
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360799 500 mg
30353648 100 mg
30362181 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30360799 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC431525500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19431-20-0
Molekylformel C27H44O
Molekylvikt (g/mol) 384.64
IUPAC-namn (3S,5S,10S,13R,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,11,12,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER CC(C)CCCC(C)[C@H]1CC=C2C3=C(CC[C@]12C)[C@@]4(C)CC[C@H](O)C[C@H]4CC3
Specifikationer
Kemiskt namn eller material (3beta,5alpha)-Cholesta-8,14-dien-3-ol
Kvantitet 500 mg
InChI formel InChI=1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h12,18-21,23,28H,6-11,13-17H2,1-5H3/t19-,20+,21+,23-,26+,27-/m1/s1
Formel vikt 384.3392