Läs mer
(3Beta)-Cholesta-4,6-dien-3-ol, TRC
Produktkod. 30012493
Change view
Kvantitet:
5 mg
30 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012493 5 mg
30009089 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012493 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2927005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14214-69-8
Molekylformel C27 H44 O
Molekylvikt (g/mol) 384.64
Synonym Cholesta-4,6-dien-3-ol, (3β)-, (3β)-Cholesta-4,6-dien-3-ol, 4,6-Cholestadien-3β-ol, Cholesta-4,6-diene-3β-ol, δ4,6-Cholestadienol
IUPAC-namn (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,8,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3C=CC4=C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Delta-4,6-cholestadienol
Kvantitet 5 mg
InChI formel InChI=1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9-10,17-19,21-25,28H,6-8,11-16H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1
Formel vikt 384.3392