Läs mer
3Beta-Hydroxy-5-cholestenoic Acid, TRC
Produktkod. 30011943
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011943 1 mg
30003291 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011943 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9175001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6561-58-6
Molekylformel C27 H44 O3
Molekylvikt (g/mol) 416.64
Synonym Cholest-5-en-26-oic acid, 3-hydroxy-, (3β)-, Cholest-5-en-26-oic acid, 3β-hydroxy- (6CI,7CI,8CI), (3β)-3-Hydroxycholest-5-en-26-oic acid, 3β-Hydroxy-5-cholestenoic acid, 3β-Hydroxycholest-5-en-26-oic acid, 3β-Hydroxycholest-5-en-27-oic acid
IUPAC-namn (6R)-6-[(3S,8S,9S,10R,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-methylheptanoic acid
LEDER C[C@H](CCCC(C)C(=O)O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 3beta-Hydroxy-5-cholestenoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C27H44O3/c1-17(6-5-7-18(2)25(29)30)22-10-11-23-21-9-8-19-16-20(28)12-14-26(19,3)24(21)13-15-27(22,23)4/h8,17-18,20-24,28H,5-7,9-16H2,1-4H3,(H,29,30)/t17-,18?,20+,21+,22-,23+,24+,26+,27-/m1/s1
Formel vikt 416.329
Procent renhet >95%