Läs mer
3Beta-Hydroxydesogestrel, TRC
Produktkod. 18063020
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18063020 10 mg
18052230 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18063020 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH93900510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70805-85-5
Molekylformel C22 H30 O2
Molekylvikt (g/mol) 326.47
Synonym 18,19-Dinorpregn-4-en-20-yne-3,17-diol, 13-ethyl-11-methylene-, (3β,17α)-, 13-Ethyl-11-methylidene-18,19-dinor-17α-pregn-4-en-20-yne-3β,17-diol, 3β-Hydroxydesogestrel, (3β,17α)-13-Ethyl-11-methylene-18,19-dinorpregn-4-en-20-yne-3,17-diol, Desogestrel Imp. E (EP)
IUPAC-namn (3S,8S,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-11-methylidene-1,2,3,6,7,8,9,10,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-diol
LEDER CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=C[C@@H](O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Specifikationer
Kemiskt namn eller material 3beta-Hydroxy-desogestrel
Kvantitet 10 mg
InChI formel InChI=1S/C22H30O2/c1-4-21-13-14(3)20-17-9-7-16(23)12-15(17)6-8-18(20)19(21)10-11-22(21,24)5-2/h2,12,16-20,23-24H,3-4,6-11,13H2,1H3/t16-,17-,18-,19-,20+,21-,22-/m0/s1
Formel vikt 326.2246