Läs mer
(3E)-1-Acetyl-3-(ethoxyphenylmethylene)-2,3-dihydro-2-oxo-1H-indole-6-carboxylic Acid Methyl Ester, TRC
Produktkod. 30323349
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30323349 50 mg
1 options
Produktkod. 30323349 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17386550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl (3E)-1-Acetyl-3-(ethoxyphenylmethylene)-2,3-dihydro-2-oxo-1H-indole-6-carboxylate
CAS 1168152-06-4
Molekylformel C21 H19 N O5
Kvantitet 50 mg
Synonym (3E)-1-Acetyl-3-(ethoxyphenylmethylene)-2,3-dihydro-2-oxo-1H-indole-6-carboxylic Acid Methyl Ester, 1H-Indole-6-carboxylic acid, 1-acetyl-3-(ethoxyphenylmethylene)-2,3-dihydro-2-oxo-, methyl ester, (3E)- (ACI), Methyl (3E)-1-acetyl-3-(ethoxyphenylmethylene)-2,3-dihydro-2-oxo-1H-indole-6-carboxylate (ACI)
InChI formel InChI=1S/C21H19NO5/c1-4-27-19(14-8-6-5-7-9-14)18-16-11-10-15(21(25)26-3)12-17(16)22(13(2)23)20(18)24/h5-12H,4H2,1-3H3/b19-18+
LEDER CCO\C(=C/1\C(=O)N(C(=O)C)c2cc(ccc12)C(=O)OC)\c3ccccc3
IUPAC-namn methyl (3E)-1-acetyl-3-[ethoxy(phenyl)methylidene]-2-oxoindole-6-carboxylate
Molekylvikt (g/mol) 365.379
Formel vikt 365.126