missing translation for 'onlineSavingsMsg'
Läs mer

(3E)-4-(Acetylthio)-Alpha-(2-chlorophenyl)-3-(2-ethoxy-2-oxoethylidene)-1-piperidineacetic Acid Methyl Ester(Mixture of Diastereomers), TRC

Produktkod. 30326928
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30326928 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30326928 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18868525MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material (3E)-4-(Acetylthio)-alpha-(2-chlorophenyl)-3-(2-ethoxy-2-oxoethylidene)-1-piperidineacetic Acid Methyl Ester(Mixture of Diastereomers)
CAS 204204-75-1
Molekylformel C20H24ClNO5S
Kvantitet 25 mg
InChI formel InChI=1S/C20H24ClNO5S/c1-4-27-18(24)11-14-12-22(10-9-17(14)28-13(2)23)19(20(25)26-3)15-7-5-6-8-16(15)21/h5-8,11,17,19H,4,9-10,12H2,1-3H3/b14-11+
LEDER CCOC(=O)\C=C\1/CN(CCC1SC(=O)C)C(C(=O)OC)c2ccccc2Cl
IUPAC-namn methyl 2-[(3E)-4-acetylsulfanyl-3-(2-ethoxy-2-oxoethylidene)piperidin-1-yl]-2-(2-chlorophenyl)acetate
Molekylvikt (g/mol) 425.93
Formel vikt 425.1064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.