Läs mer
(3E,8Z,11Z)-Tetradeca-3,8,11-trienyl Acetate (Technical Grade), TRC
Produktkod. 30135279
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135279 10 mg
30134288 100 mg
30010364 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135279 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29149310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 163041-94-9
Molekylformel C16H26O2
Molekylvikt (g/mol) 250.38
Synonym (3E,8Z,11Z)-3,8,11-Tetradecatrien-1-yl Acetate, (E,Z,Z)-3,8,11-Tetradecatrienyl Acetate;
IUPAC-namn [(3E,8Z,11Z)-tetradeca-3,8,11-trienyl] acetate
LEDER CC\C=C/C\C=C/CCC\C=C\CCOC(=O)C
Specifikationer
Kemiskt namn eller material (3E,8Z,11Z)-Tetradeca-3,8,11-trienyl Acetate
Kvantitet 10 mg
InChI formel InChI=1S/C16H26O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h4-5,7-8,12-13H,3,6,9-11,14-15H2,1-2H3/b5-4-,8-7-,13-12+
Formel vikt 250.1933