Learn More
3M™ 6000 Series Reusable Full Facepiece Respirator
Product Code. 15283467 Shop All 3M Products
Change view
Click to view available options
Format:
Medium
Unit Size:
Each
Product Code. Format Unit Size
15283467 Medium Each
1 options
Product Code. 15283467 Supplier 3M™ Supplier No. 7100015051
Quantity Price
1 1480.40 SEK / Paket med 1
4 1364.63 SEK / Paket med 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Requires
Compatible cartridges and/or filters (sold separately). Compatible with Series 2000 and 7093 filters and Series 6000 cartridges.
Requires limited maintenance time—reducing need for expensive repair programs
  • No retainers or extra parts needed to attach filters, cartridges or dual airline
  • Soft, lightweight facepiece material and easy-to-use headstraps
  • Large lens with scratch-resistant hardcoat
  • 3M Catalog# 6700 , 3M Catalog# 6800 , 3M Catalog# 6900
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Certifications
NIOSH approved for 3M Negative Pressure Air-Purifying Respirators, Positive-Pressure Dual Airline Respirators, and some 3M PAPRs (with DIN adapter).
Compliance
NIOSH approved for 3M Negative Pressure Air-Purifying Respirators, Positive-Pressure Dual Airline Respirators, and some 3M PAPRS (with DIN adapter).
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH, ANSI Z87.1-2010
Connection Type Bayonet
Material Silicone, Thermoplastic Elastomer
Product Type Full Facepiece Respirator
Format Medium
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
For Use With (Equipment) 3M™ cartridge 6000 series, filter 2000, 2200, 7000 or 5000 series or 3M™ dual airline system
Includes Large lens with scratch-resistant hard coating
Model 6800
Suspension Type 4-point