Learn More
3M™ Disposable Overboot Covers 450
The 450 disposable overboot covers from 3M are disposable accessories designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.
Brand: 3M™ 7000077618
Additional Details : Weight : 10.50000kg
Description
3M™ Overboot Covers are a reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Offered as an accessory, the overboots are made from a microporous laminate material with an anti-static treatment.
- Covers are designed with ties and elastic at the calf to help ensure a good fit
- Distinctive unisex design
- Integrated sideshields with adjustable lens angle and temple lengths
- Vertically vented browgard minimizes fogging
- 3M Catalog# 450
Specifications
Universal | |
White | |
Elastic | |
450 | |
Dry particulates, Fluid |
Microporous fabric | |
Single-use | |
Non-skid | |
Ideal to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts and for assembly, cleaning, demolition, machine operations, remediation | |
Boot Covers |