Beskrivning
- Supplied air respirators provide enhanced features compared to traditional respiratory protection.
- Offer options to cool or heat incoming air with specific valves for added comfort.
- Require a clean air source, such as plant air systems or bottled air, for operation.
- Deliver clean, breathable air to a variety of compatible hoods and helmets.
- Designed for low maintenance and reliable performance in demanding environments.
Specifikationer
Specifikationer
| Color | Black |
| Product Type | Cooling Valve Assembly |
| Size | One Size |