Läs mer
(3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride (Isopilocarpine Hydrochloride), Mikromol™
Beskrivning
(3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride (Isopilocarpine Hydrochloride)
Specifikationer
Specifikationer
| Kemiskt namn eller material | (3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride (Isopilocarpine Hydrochloride) |
| CAS | 28958-85-2 |
| Analyt- eller komponentnamn | (3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride (Isopilocarpine Hydrochloride) |
| Frakt skick | Room Temperature |
| Molekylformel | C11 H16 N2 O2 . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Pilocarpine Hydrochloride Imp. A (EP), Pilocarpine Imp. A (EP), (3R,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]dihydrofuran-2(3H)-one Hydrochloride, Pilocarpine Hydrochloride Imp. A (EP) as Hydrochloride, Isopilocarpine Hydrochloride, Pilocarpine Hydrochloride Impurity A as Hydrochloride, Pilocarpine Nitrate Impurity A as Hydrochloride, Pilocarpine Impurity A as Hydrochloride, Pilocarpine Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10+;/m0./s1 |
| LEDER | Cl.CC[C@@H]1[C@@H](Cc2cncn2C)COC1=O |
| IUPAC-namn | (3R,4R)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one;hydrochloride |
| Visa mer |