Läs mer
(3R,4R,5R)-4-(1-Ethylpropoxy)-3-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylic Acid Ethyl Ester, TRC
Produktkod. 30369190
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30369190 10 mg
1 options
Produktkod. 30369190 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE92579510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3R,4R,5R)-4-(1-Ethylpropoxy)-3-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylic acid ethyl ester
CAS 204254-94-4
Molekylformel C15 H26 O7 S
Kvantitet 10 mg
Synonym 1-Cyclohexene-1-carboxylic acid, 4-(1-ethylpropoxy)-3-hydroxy-5-[(methylsulfonyl)oxy]-, ethyl ester, (3R,4R,5R)-, 1-Cyclohexene-1-carboxylic acid, 4-(1-ethylpropoxy)-3-hydroxy-5-[(methylsulfonyl)oxy]-, ethyl ester, [3R-(3α,4α,5β)]-, (3R,4R,5R)-4-(1-Ethylpropoxy)-3-hydroxy-5-[(methylsulfonyl)oxy]-1-cyclohexene-1-carboxylic acid ethyl ester
InChI formel InChI=1S/C15H26O7S/c1-5-11(6-2)21-14-12(16)8-10(15(17)20-7-3)9-13(14)22-23(4,18)19/h8,11-14,16H,5-7,9H2,1-4H3/t12-,13-,14-/m1/s1
LEDER CCOC(=O)C1=C[C@@H](O)[C@@H](OC(CC)CC)[C@@H](C1)OS(=O)(=O)C
IUPAC-namn ethyl (3R,4R,5R)-3-hydroxy-5-methylsulfonyloxy-4-pentan-3-yloxycyclohexene-1-carboxylate
Molekylvikt (g/mol) 350.43
Formel vikt 350.1399