Learn More
(3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone, TRC
Product Code. 30407439
Change view
Click to view available options
Kvantitet:
2.5 g
Product Code. Kvantitet
30407439 2.5 g
1 options
Product Code. 30407439 Supplier Toronto Research Chemicals Supplier No. TRCH9490852.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material (3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone
CAS 132127-34-5
Molekylformel C9 H9 N O2
Kvantitet 2.5 g
InChI formel InChI=1S/C9H9NO2/c11-8-7(10-9(8)12)6-4-2-1-3-5-6/h1-5,7-8,11H,(H,10,12)/t7-,8+/m0/s1
LEDER O[C@@H]1[C@@H](NC1=O)c2ccccc2
IUPAC-namn (3R,4S)-3-hydroxy-4-phenylazetidin-2-one
Molekylvikt (g/mol) 163.17
Formel vikt 163.0633