Läs mer
(3R,5R)-6-Cyano-3,5-dihydroxy-hexanoic Acid tert-Butyl Ester, TRC
Produktkod. 30351904
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30351904 250 mg
1 options
Produktkod. 30351904 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC955901250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3R,5R)-6-Cyano-3,5-dihydroxy-hexanoic Acid tert-Butyl Ester
CAS 125971-93-9
Molekylformel C11 H19 N O4
Kvantitet 250 mg
Synonym Hexanoic acid, 6-cyano-3,5-dihydroxy-, 1,1-dimethylethyl ester, (3R,5R)-, Hexanoic acid, 6-cyano-3,5-dihydroxy-, 1,1-dimethylethyl ester, [R-(R*,R*)]-, (3R,5R)-tert-Butyl 6-cyano-3,5-dihydroxyhexanoate, (3R,5R)-tert-Butyl 6-cyano-3,5-dihydroxyhexanoate, [R-(R*,R*)]-tert-Butyl 6-cyano-3,5-dihydroxyhexanoate, tert-Butyl (3R,5R)-6-cyano-3,5-dihydroxyhexanoate
InChI formel InChI=1S/C11H19NO4/c1-11(2,3)16-10(15)7-9(14)6-8(13)4-5-12/h8-9,13-14H,4,6-7H2,1-3H3/t8-,9-/m1/s1
LEDER CC(C)(C)OC(=O)C[C@H](O)C[C@H](O)CC#N
IUPAC-namn tert-butyl (3R,5R)-6-cyano-3,5-dihydroxyhexanoate
Molekylvikt (g/mol) 229.27
Formel vikt 229.1314