Läs mer
(3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-Dimethyl-8-[[(2S)-2-methylbutanoyl]oxy]-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt (Lovastatin Hydroxy Acid Sodium Salt), Mikromol™
Beskrivning
(3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-Dimethyl-8-[[(2S)-2-methylbutanoyl]oxy]-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt (Lovastatin Hydroxy Acid Sodium Salt)
Specifikationer
Specifikationer
| Kemiskt namn eller material | (3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-Dimethyl-8-[((2S)-2-methylbutanoyl]oxy)-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt (Lovastatin Hydroxy Acid Sodium Salt) |
| CAS | 75225-50-2 |
| Analyt- eller komponentnamn | (3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-Dimethyl-8-[((2S)-2-methylbutanoyl]oxy)-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt (Lovastatin Hydroxy Acid Sodium Salt) |
| Frakt skick | Ice pack (-20°C) |
| Molekylformel | C24 H37 O6 . Na |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Lovastatin Imp. B (EP), (3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-Dimethyl-8-[[(2S)-2-methylbutanoyl]oxy]-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt, Lovastatin Imp. B (EP) as Sodium Salt, Lovastatin Hydroxy Acid Sodium Salt, Lovastatin Impurity B as Sodium Salt |
| InChI formel | InChI=1S/C24H38O6.Na/c1-5-15(3)24(29)30-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-18(25)12-19(26)13-22(27)28;/h6-7,10,14-16,18-21,23,25-26H,5,8-9,11-13H2,1-4H3,(H,27,28);/q;+1/p-1/t14-,15-,16-,18+,19+,20-,21-,23-;/m0./s1 |
| LEDER | [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12 |
| IUPAC-namn | sodium;(3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-dimethyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| Visa mer |