Läs mer
(3R,5R)-Atorvastatin Amide, TRC
Produktkod. 30135326
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135326 50 mg
30133337 100 mg
30010362 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135326 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79166550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2125501-02-0
Molekylformel C33H36FN3O4
Molekylvikt (g/mol) 557.66
Synonym 1H-Pyrrole-1-heptanamide, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, (βR,δR)-, (3R,5R)-Atorvastatin Amide, Atorvastatin Amide
IUPAC-namn 1-[(3R,5R)-7-amino-3,5-dihydroxy-7-oxoheptyl]-5-(4-fluorophenyl)-N,4-diphenyl-2-propan-2-ylpyrrole-3-carboxamide
LEDER CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)N
Specifikationer
Kemiskt namn eller material (3R,5R)-Atorvastatin Amide
Analyt- eller komponentnamn (3R,5R)-Atorvastatin Amide
Frakt skick Room Temperature
InChI formel InChI=1S/C33H36FN3O4/c1-21(2)31-30(33(41)36-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)37(31)18-17-26(38)19-27(39)20-28(35)40/h3-16,21,26-27,38-39H,17-20H2,1-2H3,(H2,35,40)(H,36,41)/t26-,27-/m1/s1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 50 mg
Formel vikt 557.269