Läs mer
(3R,5S)-6-Chloro-3,5-dihydroxyhexanoic Acid tert-Butyl Ester (~90%), TRC
Produktkod. 30365311
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30365311 1 g
1 options
Produktkod. 30365311 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3658151G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3R,5S)-6-Chloro-3,5-dihydroxyhexanoic Acid tert-Butyl Ester
CAS 154026-93-4
Molekylformel C10 H19 Cl O4
Kvantitet 1 g
Synonym Hexanoic acid, 6-chloro-3,5-dihydroxy-, 1,1-dimethylethyl ester, (3R,5S)-, Hexanoic acid, 6-chloro-3,5-dihydroxy-, 1,1-dimethylethyl ester, [S-(R*,S*)]-, 1,1-Dimethylethyl (3R,5S)-6-chloro-3,5-dihydroxyhexanoate, (3R,5S)-6-Chloro-3,5-dihydroxyhexanoic acid tert-butyl ester
InChI formel InChI=1S/C10H19ClO4/c1-10(2,3)15-9(14)5-7(12)4-8(13)6-11/h7-8,12-13H,4-6H2,1-3H3/t7-,8+/m1/s1
LEDER CC(C)(C)OC(=O)C[C@H](O)C[C@H](O)CCl
IUPAC-namn tert-butyl (3R,5S)-6-chloro-3,5-dihydroxyhexanoate
Molekylvikt (g/mol) 238.71
Formel vikt 238.0972