Läs mer
(3R,5S)-rel-5-([1,1'-Biphenyl]-4-ylmethyl)-3-methyl-2-pyrrolidinone (Racemic Mixture), TRC
Produktkod. 30417297
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417297 10 mg
30411516 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30417297 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS08102010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3R,5S)-rel-5-([1,1'-Biphenyl]-4-ylmethyl)-3-methyl-2-pyrrolidinone
CAS 2101222-51-7
Molekylformel C18 H19 N O
Kvantitet 10 mg
Synonym 2-Pyrrolidinone','5-([1,1'-biphenyl]-4-ylmethyl)-3-methyl-','(3R,5S)-rel-
InChI formel InChI=1S/C18H19NO/c1-13-11-17(19-18(13)20)12-14-7-9-16(10-8-14)15-5-3-2-4-6-15/h2-10,13,17H,11-12H2,1H3,(H,19,20)/t13-,17+/m1/s1
LEDER C[C@@H]1C[C@@H](Cc2ccc(cc2)c3ccccc3)NC1=O
IUPAC-namn (3R,5S)-3-methyl-5-[(4-phenylphenyl)methyl]pyrrolidin-2-one
Molekylvikt (g/mol) 265.35
Formel vikt 265.147