Läs mer
(3R)-Tetrahydrofuran-3-yl 4-Methylbenzenesulfonate, TRC
Produktkod. 30179786
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179786 1 g
30180242 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179786 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3102601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 219823-47-9
Molekylformel C11H14O4S
Molekylvikt (g/mol) 242.29
Synonym (R)-Toluene-4-sulfonic Acid Tetrahydrofuran-3-yl Ester, (3R)-Tetrahydro-3-furanol 4-Methylbenzenesulfonate;
IUPAC-namn [(3R)-oxolan-3-yl] 4-methylbenzenesulfonate
LEDER Cc1ccc(cc1)S(=O)(=O)O[C@@H]2CCOC2
Specifikationer
Kemiskt namn eller material (3R)-Tetrahydrofuran-3-yl 4-Methylbenzenesulfonate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C11H14O4S/c1-9-2-4-11(5-3-9)16(12,13)15-10-6-7-14-8-10/h2-5,10H,6-8H2,1H3/t10-/m1/s1
Formel vikt 242.0613
Procent renhet >95%