Läs mer
(3RS)-3-[(2-Methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-one (N-Demethylondansetron), Mikromol™
Produktkod. 30051261
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051261 25 mg
18002610 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051261 Leverantör Mikromol Leverantörsnummer MM0252.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(3RS)-3-[(2-Methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-one (N-Demethylondansetron)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3RS)-3-[(2-Methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-one (N-Demethylondansetron)
CAS 99614-14-9
Analyt- eller komponentnamn (3RS)-3-[(2-Methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-one (N-Demethylondansetron)
Frakt skick Room Temperature
Molekylformel C17 H17 N3 O
Certifikat och efterlevnad ISO 17025
Synonym Ondansetron Hydrochloride Dihydrate Imp. H (EP), Ondansetron Imp. H (EP), (3RS)-3-[(2-Methyl-1H-imidazol-1-yl)methyl]-1,2,3,9-tetrahydro-4H-carbazol-4-one, N-Demethylondansetron, Ondansetron Hydrochloride Dihydrate Impurity H, Ondansetron Impurity H
InChI formel InChI=1S/C17H17N3O/c1-11-18-8-9-20(11)10-12-6-7-15-16(17(12)21)13-4-2-3-5-14(13)19-15/h2-5,8-9,12,19H,6-7,10H2,1H3
LEDER Cc1nccn1CC2CCc3[nH]c4ccccc4c3C2=O
IUPAC-namn 3-[(2-methylimidazol-1-yl)methyl]-1,2,3,9-tetrahydrocarbazol-4-one
Molekylvikt (g/mol) 279.34
Kvantitet 25 mg
Formel vikt 279.1372 g/mol