Läs mer
(3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one, Mikromol™
Produktkod. 18021400
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18021400 100 mg
18001620 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18021400 Leverantör Mikromol Leverantörsnummer MM0252.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one
CAS 153139-56-1
Analyt- eller komponentnamn (3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one
Frakt skick Room Temperature
Molekylformel C16 H20 N2 O
Certifikat och efterlevnad ISO 17025
Synonym Ondansetron Hydrochloride Dihydrate Imp. A (EP), Ondansetron Imp. A (EP), Ondansetron USP Related Compound A, Ondansetron USP RC A, (3RS)-3-(Dimethylamino)methyl-9-methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one, Ondansetron Related Compound A, Ondansetron Hydrochloride Dihydrate Impurity A, Ondansetron Impurity A
InChI formel InChI=1S/C16H20N2O/c1-17(2)10-11-8-9-14-15(16(11)19)12-6-4-5-7-13(12)18(14)3/h4-7,11H,8-10H2,1-3H3
LEDER CN(C)CC1CCc2c(C1=O)c3ccccc3n2C
IUPAC-namn 3-[(dimethylamino)methyl]-9-methyl-2,3-dihydro-1H-carbazol-4-one
Molekylvikt (g/mol) 256.34
Kvantitet 100 mg
Formel vikt 256.1576 g/mol