Läs mer
(3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one Oxalate, Mikromol™
Beskrivning
(3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one Oxalate
Specifikationer
Specifikationer
| Kemiskt namn eller material | (3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one Oxalate |
| CAS | 1440961-11-4 |
| Analyt- eller komponentnamn | (3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one Oxalate |
| Frakt skick | Room Temperature |
| Molekylformel | C20 H19 F N2 O2 . C2 H2 O4 |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Citalopram Hydrobromide Imp. C (EP), Citalopram Imp. C (EP), Citalopram USP Related Compound C, Citalopram USP RC C, 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-3-oxo-, ethanedioate (1:1), (1RS)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-3-oxo-1,3-dihydro-2-benzofuran-5-carbonitrile oxalate, 3-Oxo Citalopram oxalate, 3-(3-N,N-Dimethylamino)-1-(4-fluorophenyl)-6-cyano-1(3H)-isobenzofuranone, (3RS)-6-Cyano-3-[3-(dimethylamino)propyl]-3-(4-fluorophenyl)isobenzofuran-1(3H)-one Oxalate, Citalopram Related Compound C (USP), Citalopram Related Compound C as Oxalate, Citalopram Hydrobromide Impurity C as Oxalate, Escitalopram Oxalate Impurity C as Oxalate, Citalopram Impurity C as Oxalate, Escitalopram Impurity C as Oxalate |
| InChI formel | InChI=1S/C20H19FN2O2.C2H2O4/c1-23(2)11-3-10-20(15-5-7-16(21)8-6-15)18-9-4-14(13-22)12-17(18)19(24)25-20;3-1(4)2(5)6/h4-9,12H,3,10-11H2,1-2H3;(H,3,4)(H,5,6) |
| LEDER | CN(C)CCCC1(OC(=O)c2cc(ccc12)C#N)c3ccc(F)cc3.OC(=O)C(=O)O |
| IUPAC-namn | 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3-oxo-2-benzofuran-5-carbonitrile;oxalic acid |
| Visa mer |