Läs mer
(3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride (2-Trifluoromethylisomer of Fluoxetine Hydrochloride), Mikromol™
Produktkod. 17942196
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17942196 100 mg
17951446 25 mg
30462565 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17942196 Leverantör Mikromol Leverantörsnummer MM0256.17
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride (2-Trifluoromethylisomer of Fluoxetine Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride
CAS 2243504-22-3
Analyt- eller komponentnamn (3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride
Frakt skick Room Temperature
Molekylformel C17 H18 F3 N O . Cl H
Certifikat och efterlevnad ISO 17025
Synonym (3RS)-N-Methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine Hydrochloride, 2-Trifluoromethylisomer of Fluoxetine Hydrochloride
InChI formel InChI=1S/C17H18F3NO.ClH/c1-21-12-11-15(13-7-3-2-4-8-13)22-16-10-6-5-9-14(16)17(18,19)20;/h2-10,15,21H,11-12H2,1H3;1H
LEDER Cl.CNCCC(Oc1ccccc1C(F)(F)F)c2ccccc2
IUPAC-namn N-methyl-3-phenyl-3-[2-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
Molekylvikt (g/mol) 345.79
Kvantitet 100 mg
Formel vikt 345.1107 g/mol