Läs mer
(3s)-1,3-Dimethylpiperazine Dihydrochloride, TRC
Produktkod. 30366647
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366647 50 mg
30355459 100 mg
30368287 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366647 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41493350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1152110-30-9
Molekylformel C6H14N2 • 2HCl
Molekylvikt (g/mol) 187.11
Synonym (S)-1,3-Dimethylpiperazine dihydrochloride
IUPAC-namn (S)-1,3-dimethylpiperazine hydrochloride
LEDER C[C@H]1CN(C)CCN1.Cl
Specifikationer
Kemiskt namn eller material (3s)-1,3-Dimethylpiperazine Dihydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C6H14N2.2ClH/c1-6-5-8(2)4-3-7-6;;/h6-7H,3-5H2,1-2H3;2*1H/t6-;;/m0../s1
Formel vikt 186.08