Läs mer
(3S,4aS,8aS)-Decahydro-N-t-butyl-3-isoquinolinecarboxamide, TRC
Produktkod. 30366597
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30366597 2.5 g 2.5g
30362818 250 mg -
30355002 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366597 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2111002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136465-81-1
Molekylformel C14 H26 N2 O
Molekylvikt (g/mol) 238.37
IUPAC-namn (3S,4aS,8aS)-N-tert-butyl-1,2,3,4,4a,5,6,7,8,8a-decahydroisoquinoline-3-carboxamide
LEDER CC(C)(C)NC(=O)[C@@H]1C[C@@H]2CCCC[C@@H]2CN1
Specifikationer
Kemiskt namn eller material (3S,4aS,8aS)-Decahydro-N-t-butyl-3-isoquinolinecarboxamide
Kvantitet 2.5 g
InChI formel InChI=1S/C14H26N2O/c1-14(2,3)16-13(17)12-8-10-6-4-5-7-11(10)9-15-12/h10-12,15H,4-9H2,1-3H3,(H,16,17)/t10-,11+,12-/m0/s1
Formel vikt 238.2045