Learn More
(3S,6S)-3,6-Dimethyl-1,4-dioxane-2,5-dione, 97%, BLD Pharm™
Product Code. 30982311
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Unit Size:
1kg
500g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30982311 100 g -
30977090 1 kg 1kg
30980752 500 g 500g
30981301 25 g -
30971696 1 g -
30981044 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30982311 Supplier BLD Pharm Supplier No. BD136525100g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 4511-42-6
Molecular Formula C6H8O4
MDL Number MFCD00070594
Quantity 100 g
SMILES O=C([C@H](C)O1)O[C@@H](C)C1=O
Molecular Weight (g/mol) 144.13
Percent Purity 97%
Chemical Name or Material (3S,6S)-3,6-Dimethyl-1,4-dioxane-2,5-dione