Läs mer
(3S-trans)-5-[[4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxyphenol (Paroxetine Metabolite), TRC
Produktkod. 17993525
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17993525 1 mg
17963705 10 mg
17964305 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17993525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5958001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 112058-89-6
Molekylformel C19 H22 F N O3
Molekylvikt (g/mol) 331.38
Synonym Paroxetine Metabolite, (3S-trans)-5-[[4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxyphenol (Paroxetine Metabolite), 4-[[(3S,4R)-4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxyphenol, (3S-trans)-4-[[4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxyphenol, BRL 36610A, Paroxetine Metabolite BRL 36583, 5-[[(3S,4R)-4-(4-Fluorophenyl)-3-piperidinyl]methoxy]-2-methoxy-phenol, BRL 36583A, (-)-trans 4-(4-Fluorophenyl)-3-(3-hydroxy-4-methoxyphenoxymethyl) piperidine, Paroxetine Metabolite BRL 36583A (Paroxetine 3-Hydroxy Metabolite)
IUPAC-namn 5-[[(3S,4R)-4-(4-fluorophenyl)piperidin-3-yl]methoxy]-2-methoxyphenol
LEDER COc1ccc(OC[C@@H]2CNCC[C@H]2c3ccc(F)cc3)cc1O
Specifikationer
Kemiskt namn eller material Paroxetine 3-Hydroxy Metabolite
Kvantitet 1 mg
InChI formel InChI=1S/C19H22FNO3/c1-23-19-7-6-16(10-18(19)22)24-12-14-11-21-9-8-17(14)13-2-4-15(20)5-3-13/h2-7,10,14,17,21-22H,8-9,11-12H2,1H3/t14-,17-/m0/s1
Formel vikt 331.1584