Läs mer
(3Z)-17-Desacetyl Norgestimate, TRC
Produktkod. 30132694
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132694 25 mg
30009794 5 mg
30007607 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132694 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28870025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
Molekylformel C21 H29 N O2
Molekylvikt (g/mol) 327.46
Specifikationer
Kemiskt namn eller material Norelgestromin (17-Deacetylnorgestimate)
CAS 53016-31-2
Kvantitet 25 mg
Synonym 18,19-Dinorpregn-4-en-20-yn-3-one, 13-ethyl-17-hydroxy-, oxime, (17α)-, 17-Deacetylnorgestimate, 18-Methylnorethindrone oxime, BRN 4202099, D-Norgestrel 3-oxime, Levonorgestrel 3-oxime, Levonorgestrel oxime, Norelgestromin, Norplant 3-oxime, RWJ 10553, d-(17α)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one oxime, 17-Desacetyl Norgestimate, Desacetylnorgestimate
InChI formel InChI=1S/C21H29NO2/c1-3-20-11-9-17-16-8-6-15(22-24)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23-24H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1
LEDER CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=NO)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
IUPAC-namn (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-3-hydroxyimino-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-ol
Formel vikt 327.2198