Läs mer
[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetonitrile, Mikromol™
Produktkod. 30461553
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461553 4 x 25 mg
17947714 100 mg
17979544 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461553 Leverantör Mikromol Leverantörsnummer MM0089.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetonitrile, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4-(1,1-Dimethylethyl)-2,6-dimethylphenyl)acetonitrile
CAS 84803-57-6
Analyt- eller komponentnamn (4-(1,1-Dimethylethyl)-2,6-dimethylphenyl)acetonitrile
Frakt skick Room Temperature
Molekylformel C14 H19 N
Certifikat och efterlevnad ISO 17025
Synonym Xylometazoline Hydrochloride Imp. C (EP), Xylometazoline Imp. C (EP), [4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetonitrile, Xylometazoline Hydrochloride Impurity C, Xylometazoline Impurity C
InChI formel InChI=1S/C14H19N/c1-10-8-12(14(3,4)5)9-11(2)13(10)6-7-15/h8-9H,6H2,1-5H3
Lösningstyp Neat
LEDER Cc1cc(cc(C)c1CC#N)C(C)(C)C
IUPAC-namn 2-(4-tert-butyl-2,6-dimethylphenyl)acetonitrile
Molekylvikt (g/mol) 201.31
Kvantitet 4 x 25 mg
Formel vikt 201.1517 g/mol