missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,3-Thiadiazol-4-yl)benzamide, 95%, Thermo Scientific™

Catalog Number p-2700363
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175205-53-5
C9H7N3OS
205.235
MFCD00173809
PAVNUEROKSNSDS-UHFFFAOYSA-N
4-1,2,3-thiadiazol-4-yl benzamide, maybridge1_007554, 4-thiadiazol-4-yl benzamide, benzamide,4-1,2,3-thiadiazol-4-yl
2739359
4-(thiadiazol-4-yl)benzamide
C1=CC(=CC=C1C2=CSN=N2)C(=O)N
This item is not returnable. View return policy
175205-53-5
C9H7N3OS
205.235
MFCD00173809
PAVNUEROKSNSDS-UHFFFAOYSA-N
4-1,2,3-thiadiazol-4-yl benzamide, maybridge1_007554, 4-thiadiazol-4-yl benzamide, benzamide,4-1,2,3-thiadiazol-4-yl
2739359
4-(thiadiazol-4-yl)benzamide
C1=CC(=CC=C1C2=CSN=N2)C(=O)N
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.24
Percent Purity 95%
Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'