Läs mer
4-(1,3,4-Oxadiazol-2-yl)aniline, TRC
Produktkod. 30427463
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427463 50 mg
30428201 10 mg
30413209 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427463 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO85862850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35219-13-7
Molekylformel C8H7N3O
Molekylvikt (g/mol) 161.16
IUPAC-namn 4-(1,3,4-oxadiazol-2-yl)aniline
LEDER Nc1ccc(cc1)c2ocnn2
Specifikationer
Kemiskt namn eller material 4-(1,3,4-Oxadiazol-2-yl)aniline
Kvantitet 50 mg
InChI formel InChI=1S/C8H7N3O/c9-7-3-1-6(2-4-7)8-11-10-5-12-8/h1-5H,9H2
Formel vikt 161.0589