Läs mer
4-[(1,3-Dihydro-1-hydroxy-2,1-benzoxaborol-5-yl)oxy]-benzamide, TRC
Produktkod. 30358066
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358066 50 mg
30353936 10 mg
30351780 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30358066 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD18859050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1187188-59-5
Molekylformel C14H12BNO4
Molekylvikt (g/mol) 269.06
Synonym Crisaborole Impurity
IUPAC-namn 4-[(1-hydroxy-3H-2,1-benzoxaborol-5-yl)oxy]benzamide
LEDER NC(=O)c1ccc(Oc2ccc3B(O)OCc3c2)cc1
Specifikationer
Kemiskt namn eller material 4-[(1,3-Dihydro-1-hydroxy-2,1-benzoxaborol-5-yl)oxy]-benzamide
Kvantitet 50 mg
InChI formel InChI=1S/C14H12BNO4/c16-14(17)9-1-3-11(4-2-9)20-12-5-6-13-10(7-12)8-19-15(13)18/h1-7,18H,8H2,(H2,16,17)
Formel vikt 269.0859