Läs mer
4'-[(1,7'-Dimethyl-2'-propyl[2,5'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxylic Acid, TRC
Produktkod. 18074030
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18074030 100 mg
18092440 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18074030 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD293045100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1026353-20-7
Molekylformel C33 H30 N4 O2
Molekylvikt (g/mol) 514.62
Synonym 4'-[(1,7'-Dimethyl-2'-propyl[2,5'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxylic acid, 4'-[(1,7'-dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazol-1'-yl)methyl]biphenyl-2-carboxylic acid, Telmisartan Imp. B (EP), Telmisartan RC B (USP)
IUPAC-namn 2-[4-[[7-methyl-5-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzoic acid
LEDER CCCc1nc2cc(cc(C)c2n1Cc3ccc(cc3)c4ccccc4C(=O)O)c5nc6ccccc6n5C
Specifikationer
Kemiskt namn eller material 4'-[(1,7'-dimethyl-2'-propyl-1H,1'H-2,5'-bibenzo[d]imidazol-1'-yl)methyl]biphenyl-2-carboxylic acid
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C33H30N4O2/c1-4-9-30-34-28-19-24(32-35-27-12-7-8-13-29(27)36(32)3)18-21(2)31(28)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19H,4,9,20H2,1-3H3,(H,38,39)
Formel vikt 514.2369
Procent renhet >95%