Learn More
4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester, 96%
Product Code. 15424766
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424766 250 mg 250mg
15414766 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15424766 Supplier Thermo Scientific Alfa Aesar Supplier No. H59075.MD
 Restricted Item
Beräknad leverans: 21-12-2026
Add to basket
CAS: 917397-94-5 | C16H19BFNO2 | 287.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 917397-94-5
Molecular Formula C16H19BFNO2
Molecular Weight (g/mol) 287.14
MDL Number MFCD22418790
InChI Key TZNIMHCBELUQIQ-UHFFFAOYSA-N
Synonym 4-1-cyanocyclopropyl-2-fluorobenzeneboronic acid pinacol ester, 1-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl cyclopropane-1-carbonitrile, 1-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl cyclopropanecarbonitrile
PubChem CID 69074412
IUPAC Name 1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carbonitrile
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C1(CC1)C#N
Specifications
Melting Point 131°C to 133°C
Quantity 250 mg
Formula Weight 287.14
Percent Purity 96%
Chemical Name or Material 4-(1-Cyanocyclopropyl)-2-fluorobenzeneboronic acid pinacol ester