Läs mer
4-(1-Methylethoxy)-N-(2-methyl-8-quinolinyl)benzamide, TRC
Produktkod. 30479399
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479399 100 mg
30467414 25 mg
30471500 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479399 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM258730100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 892711-75-0
Molekylformel C20H20N2O2
Molekylvikt (g/mol) 320.38
Synonym CDN 1163, 4-Isopropoxy-N-(2-methylquinolin-8-yl)benzamide
IUPAC-namn N-(2-methylquinolin-8-yl)-4-propan-2-yloxybenzamide
LEDER CC(C)Oc1ccc(cc1)C(=O)Nc2cccc3ccc(C)nc23
Specifikationer
Kemiskt namn eller material 4-(1-Methylethoxy)-N-(2-methyl-8-quinolinyl)benzamide
Kvantitet 100 mg
InChI formel InChI=1S/C20H20N2O2/c1-13(2)24-17-11-9-16(10-12-17)20(23)22-18-6-4-5-15-8-7-14(3)21-19(15)18/h4-13H,1-3H3,(H,22,23)
Formel vikt 320.1525