Läs mer
4-(1-Propenyl) Gemfibrozil, TRC
Produktkod. 30417493
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417493 25 mg
30413592 5 mg
30416324 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417493 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP76882025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 500904-61-0
Molekylformel C18 H26 O3
Molekylvikt (g/mol) 290.4
Synonym 5-[2,5-Dimethyl-4-(prop-1-enyl)phenoxy]-2,2-dimethylpentanoic Acid, Gemfibrozil Related Compound A (USP)
IUPAC-namn 5-(2,5-dimethyl-4-prop-1-enylphenoxy)-2,2-dimethylpentanoic acid
LEDER CC=Cc1cc(C)c(OCCCC(C)(C)C(=O)O)cc1C
Specifikationer
Kemiskt namn eller material (E,Z)-2,2-dimethyl-5-[2,5-dimethyl-4-(propene-1-yl)phenoxy]valeric acid
Kvantitet 25 mg
InChI formel InChI=1S/C18H26O3/c1-6-8-15-11-14(3)16(12-13(15)2)21-10-7-9-18(4,5)17(19)20/h6,8,11-12H,7,9-10H2,1-5H3,(H,19,20)
Formel vikt 290.1882