missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzonitrile, ≥90%, Thermo Scientific™

Catalog Number p-3110499
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
10282-30-1
C11H12N2
172.231
MFCD07368512
ZNMSYUCZLWETII-UHFFFAOYSA-N
4-pyrrolidin-1-yl benzonitrile, 4-1-pyrrolidinyl benzonitrile, 4-pyrrolidin-1-yl-benzonitrile, benzonitrile,4-1-pyrrolidinyl, 1-4-cyanophenyl pyrrolidine, 4-pyrrolidinobenzonitrile, 4-pyrrolidinylbenzonitrile, 4-pyrrolidinylbenzenecarbonitrile, 4-pyrrolidine-1-yl-benzonitrile
4961271
4-pyrrolidin-1-ylbenzonitrile
C1CCN(C1)C2=CC=C(C=C2)C#N
This item is not returnable. View return policy
10282-30-1
C11H12N2
172.231
MFCD07368512
ZNMSYUCZLWETII-UHFFFAOYSA-N
4-pyrrolidin-1-yl benzonitrile, 4-1-pyrrolidinyl benzonitrile, 4-pyrrolidin-1-yl-benzonitrile, benzonitrile,4-1-pyrrolidinyl, 1-4-cyanophenyl pyrrolidine, 4-pyrrolidinobenzonitrile, 4-pyrrolidinylbenzonitrile, 4-pyrrolidinylbenzenecarbonitrile, 4-pyrrolidine-1-yl-benzonitrile
4961271
4-pyrrolidin-1-ylbenzonitrile
C1CCN(C1)C2=CC=C(C=C2)C#N
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 172.23
Percent Purity 90%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'