missing translation for 'onlineSavingsMsg'
Läs mer

4-1BB Ligand/TNFSF9 Antibody (203942), PerCP, Novus Biologicals™

Produktkod. 30031167 Handla allt Bio Techne Produkter
Change view
Click to view available options
Kvantitet:
0.5 mL
Unit Size:
0.5ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30031167 0.5 mL 0.5ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30031167 Supplier Novus Biologicals Supplier No. FAB1246C

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Rat Monoclonal Antibody

4-1BB Ligand/TNFSF9 Monoclonal antibody specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-1BB Ligand/TNFSF9
Användningsområden Western Blot, Flow Cytometry, CyTOF
Klassificering Monoclonal
Klona 203942
Konjugera PerCP
Utspädning Western Blot, Flow Cytometry, CyTOF-ready
Formulering PBS
Gene Alias 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Värd art Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse 4-1BB Ligand/TNFSF9, Arg104-Glu309, Accession # P41274
Reningsmetod Protein A or G purified from hybridoma culture supernatant
Kvantitet 0.5 mL
Regulatorisk status RUO
Forskningsdisciplin Apoptosis, Biologically Active Proteins
Primär eller sekundär Primary
Gen-ID (Entrez) 8744
Målarter Mouse
Innehåll och lagring Store at 4C in the dark.
Form Purified
Isotyp IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.