missing translation for 'onlineSavingsMsg'
Learn More

[4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol 97+%, Thermo Scientific™

Catalog Number p-4510726
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
160388-55-6
C11H12N2O
188.23
MFCD08435916
ADLULGOPZPFWPA-UHFFFAOYSA-N
4-1h-pyrazol-1-ylmethyl phenyl methanol, 4-pyrazol-1-ylmethyl phenyl methanol, 4-1h-pyrazol-1-yl methyl phenyl methanol, 1-4-hydroxymethyl-benzyl-1h-pyrazole, benzenemethanol, 4-1h-pyrazol-1-ylmethyl, 9vq
16640562
[4-(pyrazol-1-ylmethyl)phenyl]methanol
OCC1=CC=C(CN2C=CC=N2)C=C1
This item is not returnable. View return policy
160388-55-6
C11H12N2O
188.23
MFCD08435916
ADLULGOPZPFWPA-UHFFFAOYSA-N
4-1h-pyrazol-1-ylmethyl phenyl methanol, 4-pyrazol-1-ylmethyl phenyl methanol, 4-1h-pyrazol-1-yl methyl phenyl methanol, 1-4-hydroxymethyl-benzyl-1h-pyrazole, benzenemethanol, 4-1h-pyrazol-1-ylmethyl, 9vq
16640562
[4-(pyrazol-1-ylmethyl)phenyl]methanol
OCC1=CC=C(CN2C=CC=N2)C=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material [4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'