missing translation for 'onlineSavingsMsg'
Learn More

4(1H)-Pyrimidinone, 98%, Thermo Scientific Chemicals

Catalog Number 15451999
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4562-27-0
C4H4N2O
96.089
MFCD00006664
DNCYBUMDUBHIJZ-UHFFFAOYSA-N
pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine
20695
1H-pyrimidin-6-one
C1=CN=CNC1=O
This item is not returnable. View return policy
4562-27-0
C4H4N2O
96.089
MFCD00006664
DNCYBUMDUBHIJZ-UHFFFAOYSA-N
pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine
20695
1H-pyrimidin-6-one
C1=CN=CNC1=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 163°C to 169°C (literature)
Quantity 1 g
Beilstein 741972
Formula Weight 96.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4(1H)-Pyrimidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'