Learn More
4-(1H-Pyrrol-1-yl)benzaldehyde, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 23351-05-5 |
| Molecular Formula | C11H9NO |
| MDL Number | MFCD03822126 |
| Quantity | 100 mg |
| SMILES | O=CC1=CC=C(C=C1)N1C=CC=C1 |
| Molecular Weight (g/mol) | 171.20 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(1H-Pyrrol-1-yl)benzaldehyde |