Läs mer
4-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol, Mikromol™
Produktkod. 30049287
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049287 25 mg
17971096 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049287 Leverantör Mikromol Leverantörsnummer MM1122.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol
CAS 949095-98-1
Analyt- eller komponentnamn 4-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol
Frakt skick Room Temperature
Molekylformel C18 H19 N O S
Certifikat och efterlevnad ISO 9001
Synonym 4-Bromobenzhydrol, Duloxetine Hydrochloride Imp. C (EP), Duloxetine Imp. C (EP), 1-Naphthalenol, 4-[3-(methylamino)-1-(2-thienyl)propyl]-, 4-[3-(Methylamino)-1-(2-thienyl)propyl]-1-naphthalenol, Para-Naphthol duloxetine, 4-[(1RS)-3-(Methylamino)-1-(thiophen-2-yl)propyl]naphthalen-1-ol, Duloxetine Hydrochloride Impurity C, Duloxetine Impurity C
LEDER CNCCC(c1cccs1)c2ccc(O)c3ccccc23
Molekylvikt (g/mol) 297.42
Kvantitet 25 mg
Formel vikt 297.416 g/mol