Läs mer
4-[2-(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)ethoxy]-3-oxobutanoic Acid Methyl Ester, TRC
Produktkod. 30374339
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30374339 50 mg
1 options
Produktkod. 30374339 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45011550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[2-(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)ethoxy]-3-oxobutanoic Acid Methyl Ester
CAS 111429-90-4
Molekylformel C15H15NO6
Kvantitet 50 mg
Synonym 4-[2-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)ethoxy]-3-oxobutyric Acid Methyl Ester
InChI formel InChI=1S/C15H15NO6/c1-21-13(18)8-10(17)9-22-7-6-16-14(19)11-4-2-3-5-12(11)15(16)20/h2-5H,6-9H2,1H3
LEDER O=C1N(CCOCC(CC(OC)=O)=O)C(C2=CC=CC=C21)=O
IUPAC-namn methyl 4-(2-(1,3-dioxoisoindolin-2-yl)ethoxy)-3-oxobutanoate
Molekylvikt (g/mol) 305.28
Formel vikt 305.09