Läs mer
4-[2-(1-Piperidinyl)ethoxy]benzoic Acid Hydrochloride Salt, TRC
Produktkod. 30424368
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30424368 25 mg
1 options
Produktkod. 30424368 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP48202525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[2-(1-Piperidinyl)ethoxy]benzoic Acid Hydrochloride
CAS 84449-80-9
Molekylformel C14 H19 N O3 . Cl H
Kvantitet 25 mg
Synonym Benzoic acid','4-[2-(1-piperidinyl)ethoxy]-','hydrochloride (1:1), Benzoic acid','4-[2-(1-piperidinyl)ethoxy]-','hydrochloride (9CI), 4-(2-Piperidinoethoxy)benzoic acid hydrochloride
InChI formel InChI=1S/C14H19NO3.ClH/c16-14(17)12-4-6-13(7-5-12)18-11-10-15-8-2-1-3-9-15;/h4-7H,1-3,8-11H2,(H,16,17);1H
LEDER Cl.OC(=O)c1ccc(OCCN2CCCCC2)cc1
IUPAC-namn 4-(2-piperidin-1-ylethoxy)benzoic acid;hydrochloride
Molekylvikt (g/mol) 285.77
Formel vikt 285.1132