Läs mer
4-[2,2,2-Trifluoro-1-[4-(phenylmethoxy)phenyl]-1-(trifluoromethyl)ethyl]-phenol, TRC
Produktkod. 30450897
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450897 25 mg
30450009 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30450897 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT79277025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[2,2,2-Trifluoro-1-[4-(phenylmethoxy)phenyl]-1-(trifluoromethyl)ethyl]-phenol
CAS 685561-26-6
Molekylformel C22H16F6O2
Kvantitet 25 mg
InChI formel InChI=1S/C22H16F6O2/c23-21(24,25)20(22(26,27)28,16-6-10-18(29)11-7-16)17-8-12-19(13-9-17)30-14-15-4-2-1-3-5-15/h1-13,29H,14H2
LEDER Oc1ccc(cc1)C(c2ccc(OCc3ccccc3)cc2)(C(F)(F)F)C(F)(F)F
IUPAC-namn 4-[1,1,1,3,3,3-hexafluoro-2-(4-phenylmethoxyphenyl)propan-2-yl]phenol
Molekylvikt (g/mol) 426.35
Formel vikt 426.1054